Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7-Dibromo-2,1,3-benzothiadiazole | 15155-41-6 | 293.97 | 250 G
SDP

Catalog No. 5000450051
Encompass_Preferred

4,7-Dibromo-2,1,3-benzothiadiazole is a biochemical reagent, appearing as a light brown to yellow solid, intended for life science related research. This product is for research use only and is not sold to patients.

  • Can be used as a biological material or organic compound
  • Appearance: solid
  • Color: light brown to yellow
  • In vitro solubility in DMSO: 25 mg/mL (85.04 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-50051 Supplier Medchemexpress LLC Supplier No. 250GHYW002651
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4,7-Dibromo-2,1,3-benzothiadiazole is a biochemical reagent, appearing as a light brown to yellow solid, intended for life science related research. This product is for research use only and is not sold to patients.

  • Can be used as a biological material or organic compound
  • Appearance: solid
  • Color: light brown to yellow
  • In vitro solubility in DMSO: 25 mg/mL (85.04 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.