Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,7-Dibromo-2,1,3-benzothiadiazole | 15155-41-6 | 293.97 | 50 G
SDP

Catalog No. 5000448687
Encompass_Preferred

4,7-Dibromo-2,1,3-benzothiadiazole is a biochemical reagent intended for life science related research. It can be utilized as a biological material or organic compound and is for research use only.

  • Molecular weight of 293.97
  • Chemical formula C6H2Br2N2S
  • CAS number: 15155-41-6
  • Appears as a light brown to yellow solid
  • Purity of 99.89%
  • Soluble in DMSO (25 mg/mL) and corn oil for in vivo applications
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-48687 Supplier Medchemexpress LLC Supplier No. HYW00265150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4,7-Dibromo-2,1,3-benzothiadiazole is a biochemical reagent intended for life science related research. It can be utilized as a biological material or organic compound and is for research use only.

  • Molecular weight of 293.97
  • Chemical formula C6H2Br2N2S
  • CAS number: 15155-41-6
  • Appears as a light brown to yellow solid
  • Purity of 99.89%
  • Soluble in DMSO (25 mg/mL) and corn oil for in vivo applications
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.