Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole 98.0+%, TCI America™
SDP

Catalog No. D4734200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4734200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4734200MG Supplier TCI America Supplier No. D4734200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1295502-53-2
Molecular Formula C6Br2F2N2S
Molecular Weight (g/mol) 329.945
InChI Key HFUBKQHDPJZQIW-UHFFFAOYSA-N
PubChem CID 59523104
IUPAC Name 4,7-dibromo-5,6-difluoro-2,1,3-benzothiadiazole
SMILES C1(=C(C2=NSN=C2C(=C1F)Br)Br)F

Specifications

Melting Point 155°C
Color White
Quantity 200 mg
Formula Weight 329.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.