Learn More
Medchemexpress LLC 4,7-dihydro-6-(1-methylethyl)-7-oxo-5-phenyl-pyrazolo[1,5-a]pyrimidine-3-carbonitrile | 1628208-23-0 | 99.0% | 278.31 g/mol | C16H14N4O | 1 MG
SDP
Catalog No. 5000214795
Encompass_Preferred
CPI-455 is a potent, cell-active pan-KDM5 (histone lysine demethylase 5 family) inhibitor used in research to increase global H3K4 trimethylation and investigate KDM5-dependent biology. It is supplied as a white to off-white solid and has documented enzymatic potency and analytical characterization suitable for laboratory studies.
  • Potent pan-KDM5 inhibition with low-nanomolar biochemical activity.
  • Promotes increased H3K4 trimethylation in cellular models.
  • High purity appropriate for research applications.
  • Solid form for convenient handling and storage.
  • Documented analytical data and supporting datasheet available.
Catalog No. 50-002-14795 Supplier Medchemexpress LLC Supplier No. HY1004211MG
CPI-455 is a potent, cell-active pan-KDM5 (histone lysine demethylase 5 family) inhibitor used in research to increase global H3K4 trimethylation and investigate KDM5-dependent biology. It is supplied as a white to off-white solid and has documented enzymatic potency and analytical characterization suitable for laboratory studies.
  • Potent pan-KDM5 inhibition with low-nanomolar biochemical activity.
  • Promotes increased H3K4 trimethylation in cellular models.
  • High purity appropriate for research applications.
  • Solid form for convenient handling and storage.
  • Documented analytical data and supporting datasheet available.