Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dihydroxy-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. D38691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38691G Supplier TCI America Supplier No. D38691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3922-40-5
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.208
MDL Number MFCD00004975
InChI Key SLIBCJURSADKPV-UHFFFAOYSA-N
Synonym 1,10-phenanthroline-4,7-diol, 4,7-dihydroxy-1,10-phenanthroline, 1,10-phenanthroline-4,7 1h,10h-dione, 1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione, pyridino 3,2-h quinoline-4,7-diol, acmc-1coeh, bidd:gt0532, 1,1-phenanthroline-4,7-diol, 1,10 phenanthroline-4,7-diol, 4,7-dioxylato-1,10-phenanthroline
PubChem CID 77524
IUPAC Name 1,10-dihydro-1,10-phenanthroline-4,7-dione
SMILES C1=CC2=C(C3=C1C(=O)C=CN3)NC=CC2=O

Specifications

Quantity 1 g
Formula Weight 212.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dihydroxy-1,10-phenanthroline
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.