Learn More
Medchemexpress LLC 4',7-Dimethoxyisoflavone | 1157-39-7 | 10 MM 1 ML
SDP
Catalog No. 5000421767
Encompass_Preferred
4',7-Dimethoxyisoflavone, also known as Dimethoxydaidzein, is a natural compound isolated from the leaves of Albizzia lebbeck. It exhibits antifungal activity against various plant pathogenic fungi and is intended for research use only.
  • Isolated from Albizzia lebbeck
  • Exhibits antifungal activity
  • For research use only
  • High purity (98.20%)
  • White to off-white solid appearance
  • Soluble in DMSO
Catalog No. 50-004-21767 Supplier Medchemexpress LLC Supplier No. HYN214510MM/1ML
4',7-Dimethoxyisoflavone, also known as Dimethoxydaidzein, is a natural compound isolated from the leaves of Albizzia lebbeck. It exhibits antifungal activity against various plant pathogenic fungi and is intended for research use only.
  • Isolated from Albizzia lebbeck
  • Exhibits antifungal activity
  • For research use only
  • High purity (98.20%)
  • White to off-white solid appearance
  • Soluble in DMSO