Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4',7-Dimethoxyisoflavone | 1157-39-7 | 25 MG
SDP

Catalog No. 5000420714
Encompass_Preferred

4',7-Dimethoxyisoflavone is isolated from the leaves of *Albizzia lebbeck* and demonstrates antifungal activity. In vitro studies indicate its effectiveness against various plant pathogenic fungi, with sensitivity varying among different fungal species.

  • Purity: 98.20%
  • Molecular weight: 282.29
  • Formula: C17H14O4
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Flavonoids, isoflavones
  • Initial source: Plants, Leguminosae, *Glycine max (L.) merr*
  • Endogenous metabolite: Yes

Catalog No. 50-004-20714 Supplier Medchemexpress LLC Supplier No. 25MGHYN2145
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4',7-Dimethoxyisoflavone is isolated from the leaves of *Albizzia lebbeck* and demonstrates antifungal activity. In vitro studies indicate its effectiveness against various plant pathogenic fungi, with sensitivity varying among different fungal species.

  • Purity: 98.20%
  • Molecular weight: 282.29
  • Formula: C17H14O4
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Flavonoids, isoflavones
  • Initial source: Plants, Leguminosae, *Glycine max (L.) merr*
  • Endogenous metabolite: Yes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.