Learn More
Strem, An Ascensus Company CAS# 1662-01-7. 1g. 4,7-Diphenyl-1,10-phenanthroline, 99% (Bathophenanthroline). MFCD00004976
SDP
Catalog No. 5090109003
Encompass
CAS# 1662-01-7. 1g. 4,7-Diphenyl-1,10-phenanthroline, 99% (Bathophenanthroline). MFCD00004976. Molecular Weight: 332.40. Molecular Formula: C24H16N2. Color/form: off-white pwdr. Strem# 07-0472. http://www.strem.com/catalog/v/07-0472/
Catalog No. 50-901-09003 Supplier Strem, An Ascensus Company Supplier No. 0704721G
CAS# 1662-01-7. 1g. 4,7-Diphenyl-1,10-phenanthroline, 99% (Bathophenanthroline). MFCD00004976. Molecular Weight: 332.40. Molecular Formula: C24H16N2. Color/form: off-white pwdr. Strem# 07-0472. http://www.strem.com/catalog/v/07-0472/