Learn More
Quality Lab Accessories 4.75IN BENT 316 SS CANNULA
Catalog No. NC2542864
Encompass
4.75'' (120mm) Bent 316 SS Cannula with SS Luer Lock & Perm SS Tip for Hanson/QLA "HR" style filters. For 900mL Sampling.
Catalog No. NC2542864 Supplier Quality Lab Accessories Supplier No. CAN475HR
4.75'' (120mm) Bent 316 SS Cannula with SS Luer Lock & Perm SS Tip for Hanson/QLA "HR" style filters. For 900mL Sampling.