Learn More
Quality Lab Accessories 4.75" (120mm) Bent 316 SS 1/8" (3.2mm) OD Cannula with SS Luer Lock & PTFE sleeve for QLA "01" style filters
Catalog No. NC9161955
Encompass
4.75" (120mm) Bent 316 SS 1/8" (3.2mm) OD Cannula with SS Luer Lock & PTFE sleeve for QLA "01" style filters
Catalog No. NC9161955 Supplier Quality Lab Accessories Supplier No. CA8475VK
4.75" (120mm) Bent 316 SS 1/8" (3.2mm) OD Cannula with SS Luer Lock & PTFE sleeve for QLA "01" style filters