Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,8-Bis(hydroxymethyl)tricyclo[5.2.1.02,6]decane, mixture of isomers

Catalog No. B45909500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45909500G 500 g
B45909100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45909500G Supplier Sigma Aldrich Supplier No. B45909500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4,8-bis(hydroxymethyl)tricyclo[5.2.1.02,6]decane (tdmol) is used in the preparation of high molecular weight poly(1,4-butylene carbonate) (pbc)
  • It is also used in the synthesis of photoactive compounds (pac) for the preparation of positive photo-resist materials

Specifications

CAS 26896-48-0
Molecular Formula C12H20O2
Refractive Index n20/D 1.528 (literature)
Linear Formula C12H20O2
MDL Number MFCD00151166
Quantity 500 g
Synonym TDMol; Tricyclo[5.2.1.02,6 ]decane-4,8-dimethanol
Molecular Weight (g/mol) 196.29
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.