Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde 97.0+%, TCI America™
SDP

Catalog No. B5484200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5484200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5484200MG Supplier TCI America Supplier No. B5484200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1668554-22-0
Molecular Formula C28H38O4S2
Molecular Weight (g/mol) 502.728
InChI Key JDPODAXHWNAIOO-UHFFFAOYSA-N
PubChem CID 122234142
IUPAC Name 4,8-dioctoxythieno[2,3-f][1]benzothiole-2,6-dicarbaldehyde
SMILES CCCCCCCCOC1=C2C=C(SC2=C(C3=C1SC(=C3)C=O)OCCCCCCCC)C=O

Specifications

Melting Point 106°C
Quantity 200 mg
Formula Weight 502.73
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b′]dithiophene-2,6-dicarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.