Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,8-Dihydroxyquinoline-2-carboxylic acid, 96%

Catalog No. AC115090010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC115090010

Supplier: Thermo Scientific Chemicals 115090010

Only null left
Add to Cart

CAS: 59-00-7 | C10H7NO4 | 205.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-00-7
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.17
MDL Number MFCD00006754
InChI Key FBZONXHGGPHHIY-UHFFFAOYSA-N
Synonym xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
PubChem CID 5699
ChEBI CHEBI:10072
SMILES OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 297°C to 298°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10266
IUPAC Name 8-hydroxy-4-oxo-1H-quinoline-2-carboxylic acid
Formula Weight 205.17
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4, 8-Dihydroxyquinoline-2-carboxylic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-410-1
RTECSNumber UZ9275000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.