Learn More
Cayman Chemical 4 8-Iso-THC 100ug
SDP
Catalog No. 502947365
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; an impurity in the conversion of cannabidiol to Δ8-THC or Δ9-THC; intended for research and forensic applications
Catalog No. 50-294-7365 Supplier Cayman Chemical Supplier No. 33863100 UG
An analytical reference standard that is structurally similar to known phytocannabinoids; an impurity in the conversion of cannabidiol to Δ8-THC or Δ9-THC; intended for research and forensic applications