Learn More
Medchemexpress LLC 4,8-Methanobenzofuro[2,3-a]pyrido[4,3-b]carbazole-12-carboxylic acid, 7-(cyclopropylmethyl)-5,6,7,8,8a,9,14,14b-octahydro-1,8a-dihydroxy-, ethyl ester, (4bS,8R,8aS,14bR)- | 892039-23-5 | 99.44% | 486.56 | 100 MG
SDP
Catalog No. 5000350201
Encompass_Preferred
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist with a Ki of 0.47 nM and a Kb of 0.21 nM. It also acts as an antagonist for μ-opioid receptor (MOR; Ki=36.56 nM and Kb=9.43 nM) and κ-opioid receptor (KOR; Ki=5.31 nM and Kb=7.18 nM). TAN-452, a derivative of Naltrindole, exhibits low brain penetrability and can attenuate morphine-induced side effects without impacting pain control.
  • Orally active
  • Selective peripherally acting δ-opioid receptor (DOR) antagonist
  • Low brain penetrability
  • Attenuates morphine-induced side effects without affecting pain control
  • Antagonist for μ-opioid receptor (MOR) and κ-opioid receptor (KOR)
Catalog No. 50-003-50201 Supplier Medchemexpress LLC Supplier No. HY136208100MG
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist with a Ki of 0.47 nM and a Kb of 0.21 nM. It also acts as an antagonist for μ-opioid receptor (MOR; Ki=36.56 nM and Kb=9.43 nM) and κ-opioid receptor (KOR; Ki=5.31 nM and Kb=7.18 nM). TAN-452, a derivative of Naltrindole, exhibits low brain penetrability and can attenuate morphine-induced side effects without impacting pain control.
  • Orally active
  • Selective peripherally acting δ-opioid receptor (DOR) antagonist
  • Low brain penetrability
  • Attenuates morphine-induced side effects without affecting pain control
  • Antagonist for μ-opioid receptor (MOR) and κ-opioid receptor (KOR)