Learn More
Medchemexpress LLC 4,8-Methanobenzofuro[2,3-a]pyrido[4,3-b]carbazole-12-carboxylic acid, 7-(cyclopropylmethyl)-5,6,7,8,8a,9,14,14b-octahydro-1,8a-dihydroxy-, ethyl ester, (4bS,8R,8aS,14bR)- | 892039-23-5 | 99.44% | 486.56 | 25 MG
SDP
Catalog No. 5000351579
Encompass_Preferred
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist. It exhibits low brain penetrability and can alleviate morphine-induced side effects without affecting pain control.
  • Orally active and selective δ-opioid receptor (DOR) antagonist
  • Low brain penetrability
  • Alleviates morphine-induced side effects without impacting pain control
  • Also acts as an antagonist for μ-opioid and κ-opioid receptors
Catalog No. 50-003-51579 Supplier Medchemexpress LLC Supplier No. 25MGHY136208
TAN-452 is an orally active, selective peripherally acting δ-opioid receptor (DOR) antagonist. It exhibits low brain penetrability and can alleviate morphine-induced side effects without affecting pain control.
  • Orally active and selective δ-opioid receptor (DOR) antagonist
  • Low brain penetrability
  • Alleviates morphine-induced side effects without impacting pain control
  • Also acts as an antagonist for μ-opioid and κ-opioid receptors