Learn More
Cayman Chemical 4 8DIoxa3HperfluoronnoI 10mg
SDP
Catalog No. 502917687
Encompass_Preferred
A diterpene alkaloid inhibits Nav1.7 in HEK293 cells expressing the human channel (IC50 25.8 nM) scavenges DPPH radicals by 65.3 at 1 mM increases the mechanical pain threshold in the von Frey test in streptozotocin-induced diabetic but not non-diabetic mice at 10 mg/kg
Catalog No. 50-291-7687 Supplier Cayman Chemical Supplier No. 3726710 MG
A diterpene alkaloid inhibits Nav1.7 in HEK293 cells expressing the human channel (IC50 25.8 nM) scavenges DPPH radicals by 65.3 at 1 mM increases the mechanical pain threshold in the von Frey test in streptozotocin-induced diabetic but not non-diabetic mice at 10 mg/kg