Learn More
Fine Science Tools Instrument Tip Protectors, Clear, 4.8 x 25.4 mm
SDP
Catalog No. NC9388461
Encompass
Product: Instrument Tip Protectors - 4.8 x 25.4mm/Clear/ Dimensions: 4.8 x 25.4mm/ Color: Clear/ Material: Plastisol Autoclave Safe: Yes/ Unit: 25
Catalog No. NC9388461 Supplier Fine Science Tools Supplier No. 2906005
Product: Instrument Tip Protectors - 4.8 x 25.4mm/Clear/ Dimensions: 4.8 x 25.4mm/ Color: Clear/ Material: Plastisol Autoclave Safe: Yes/ Unit: 25