Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(9-Carbazolyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog No. AAH5194803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5194803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5194803 Supplier Thermo Scientific Chemicals Supplier No. H5194803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 785051-54-9
Molecular Formula C24H24BNO2
Molecular Weight (g/mol) 369.271
MDL Number MFCD16294549
InChI Key AHDSYMVAUJZCOP-UHFFFAOYSA-N
Synonym 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbazole, amtb250, 9-carbazolylbenzeneboronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid pinacol ester, 9h-carbazole-9-4-phenyl boronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid, pinacol ester
PubChem CID 11850073
IUPAC Name 9-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53

Specifications

Quantity 1 g
Beilstein 10465891
Sensitivity Air Sensitive
Formula Weight 369.27
Percent Purity 95%
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.