Learn More
Cayman Chemical 4 9-nhydrotetrodotoxIn 100ug
SDP
Catalog No. 502981898
Encompass_Preferred
A selective IDO1 inhibitor (IC50s 67 and 19 nM for human IDO in enzymatic activity and HeLa cell assays respectively) decreases kynurenine levels in plasma and dose-dependently reduces the growth of GM-CSF-secreting B16 tumor xenografts in mice at 25-75 mg/kg
Catalog No. 50-298-1898 Supplier Cayman Chemical Supplier No. 19084100 UG
A selective IDO1 inhibitor (IC50s 67 and 19 nM for human IDO in enzymatic activity and HeLa cell assays respectively) decreases kynurenine levels in plasma and dose-dependently reduces the growth of GM-CSF-secreting B16 tumor xenografts in mice at 25-75 mg/kg