Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. F1050200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1050200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1050200MG Supplier TCI America Supplier No. F1050200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 204320-51-4
Molecular Formula C20H19NO5
Molecular Weight (g/mol) 353.374
MDL Number MFCD01632020
InChI Key CJVIYWXADATNKP-UHFFFAOYSA-N
Synonym 4-Fmoc-morpholine-3-carboxylic Acid
PubChem CID 4461441
IUPAC Name 4-(9H-fluoren-9-ylmethoxycarbonyl)morpholine-3-carboxylic acid
SMILES C1COCC(N1C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 353.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.