Learn More
Cayman Chemical Δ4-Abiraterone, Purity: ≥98%, CAS: 154229-21-7, 10 mg
SDP
Catalog No. 502938679
Encompass_Preferred
An active metabolite of abiraterone; androgen receptor antagonist (IC50 = 5.3 nM); reduces expression of androgen receptor target genes in LNCaP, LAPC4, and C4-2 cell lines; inhibits CYP17A1 at 1 nM; blocks SRD5A conversion of Δ4-androstenedione to 5α-androstanedione in LAPC4 cells at 10 μM; delays tumor progression in a VCaP mouse xenograft model; increases progression-free survival in a C4-2 mouse xenograft model
Catalog No. 50-293-8679 Supplier Cayman Chemical Supplier No. 2759410 MG
An active metabolite of abiraterone; androgen receptor antagonist (IC50 = 5.3 nM); reduces expression of androgen receptor target genes in LNCaP, LAPC4, and C4-2 cell lines; inhibits CYP17A1 at 1 nM; blocks SRD5A conversion of Δ4-androstenedione to 5α-androstanedione in LAPC4 cells at 10 μM; delays tumor progression in a VCaP mouse xenograft model; increases progression-free survival in a C4-2 mouse xenograft model