Learn More
Medchemexpress LLC 4-acetamido-4'-isothiocyanato-2,2'-stilbenedisulfonic acid disodium salt hydrate | 51023-76-8 | MFCD00007485 | ≥80.0% | 498.46 g·mol-1 (anhydrous basis) | C17H12N2Na2O7S3 · xH2O | 5 MG
SDP
Catalog No. 5000222433
Encompass_Preferred
4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium (SITS) is the disodium salt of an isothiocyanate-functionalized stilbenedisulfonate used as a biochemical reagent. It is employed as a labeling reagent and an anion transport inhibitor in cell and membrane studies and is supplied as a hydrated disodium salt.
  • Reactive isothiocyanate group for covalent labeling of proteins and amines.
  • Useful probe for studying anion transport and membrane permeability.
  • Water-soluble disodium salt suitable for aqueous biological buffers.
  • Available in small research quantities for experimental assays.
  • Purity typically ≥80% as specified by suppliers.
Catalog No. 50-002-22433 Supplier Medchemexpress LLC Supplier No. HY1378965MG
4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium (SITS) is the disodium salt of an isothiocyanate-functionalized stilbenedisulfonate used as a biochemical reagent. It is employed as a labeling reagent and an anion transport inhibitor in cell and membrane studies and is supplied as a hydrated disodium salt.
  • Reactive isothiocyanate group for covalent labeling of proteins and amines.
  • Useful probe for studying anion transport and membrane permeability.
  • Water-soluble disodium salt suitable for aqueous biological buffers.
  • Available in small research quantities for experimental assays.
  • Purity typically ≥80% as specified by suppliers.