Learn More
Medchemexpress LLC 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium | 51023-76-8 | 94.37% | 498.46 | 25 MG
SDP
Catalog No. 5000348946
Encompass_Preferred
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent dye. It can be utilized to demonstrate retrograde axonal transport, to label secondary antibodies, and as a fluorescent whole cell stain. This product is for research use only.
  • Can be utilized to demonstrate retrograde axonal transport
  • Labels secondary antibodies
  • Functions as a fluorescent whole cell stain
Catalog No. 50-003-48946 Supplier Medchemexpress LLC Supplier No. 25MGHY137896
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent dye. It can be utilized to demonstrate retrograde axonal transport, to label secondary antibodies, and as a fluorescent whole cell stain. This product is for research use only.
  • Can be utilized to demonstrate retrograde axonal transport
  • Labels secondary antibodies
  • Functions as a fluorescent whole cell stain