Learn More
Medchemexpress LLC 4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium | 51023-76-8 | 94.37% | 498.46 | 50 MG
SDP
Catalog No. 5000350705
Encompass_Preferred
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent dye that has a purity of 94.37%. It appears as a light yellow to yellow solid. This compound is intended for research use only and is not sold to patients.
  • Functions as a fluorescent dye.
  • Can be utilized to demonstrate retrograde axonal transport.
  • Suitable for labeling secondary antibodies.
  • Can be used as a fluorescent whole cell stain.
  • Soluble in DMSO at 250 mg/mL (501.54 mM).
  • Should be stored at -20°C, protected from light, and under nitrogen. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month (protected from light, stored under nitrogen).
Catalog No. 50-003-50705 Supplier Medchemexpress LLC Supplier No. HY13789650MG
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent dye that has a purity of 94.37%. It appears as a light yellow to yellow solid. This compound is intended for research use only and is not sold to patients.
  • Functions as a fluorescent dye.
  • Can be utilized to demonstrate retrograde axonal transport.
  • Suitable for labeling secondary antibodies.
  • Can be used as a fluorescent whole cell stain.
  • Soluble in DMSO at 250 mg/mL (501.54 mM).
  • Should be stored at -20°C, protected from light, and under nitrogen. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month (protected from light, stored under nitrogen).