Learn More
Medchemexpress LLC 4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (disodium) | 51023-76-8 | 94.4% | 498.46 | 100 MG
SDP
Catalog No. 5000351219
Encompass_Preferred
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (disodium) is a fluorescent dye. It is utilized in various biological applications.
  • Demonstrates retrograde axonal transport
  • Labels secondary antibodies
  • Used as a fluorescent whole cell stain
Catalog No. 50-003-51219 Supplier Medchemexpress LLC Supplier No. HY137896100MG
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (disodium) is a fluorescent dye. It is utilized in various biological applications.
  • Demonstrates retrograde axonal transport
  • Labels secondary antibodies
  • Used as a fluorescent whole cell stain