Learn More
Medchemexpress LLC 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium | 51023-76-8 | MFCD00007485 | 94.4% | C17H12N2Na2O7S3 | 10MG
SDP
Catalog No. 5000259845
Encompass_Preferred
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent stilbene-derived dye and membrane probe used in cellular labeling and transport studies. It reacts covalently with proteins via its isothiocyanate group and is applied to visualize retrograde axonal transport, label antibodies, and stain whole cells; it also modulates anion permeability and Na,K-ATPase activity.
  • Fluorescent stilbene dye for cell and membrane staining.
  • Isothiocyanate group enables covalent labeling of proteins.
  • Suitable for demonstrating retrograde axonal transport.
  • Useful for studying anion transport and Na,K-ATPase activity.
  • Provided as a water-soluble disodium salt for biological use.
  • Available in small research pack sizes for laboratory studies.
Catalog No. 50-002-59845 Supplier Medchemexpress LLC Supplier No. HY13789610MG
4-Acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid disodium is a fluorescent stilbene-derived dye and membrane probe used in cellular labeling and transport studies. It reacts covalently with proteins via its isothiocyanate group and is applied to visualize retrograde axonal transport, label antibodies, and stain whole cells; it also modulates anion permeability and Na,K-ATPase activity.
  • Fluorescent stilbene dye for cell and membrane staining.
  • Isothiocyanate group enables covalent labeling of proteins.
  • Suitable for demonstrating retrograde axonal transport.
  • Useful for studying anion transport and Na,K-ATPase activity.
  • Provided as a water-soluble disodium salt for biological use.
  • Available in small research pack sizes for laboratory studies.