Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-acetamido-5-chloro-2-methoxybenzoic acid | 24201-13-6 | 99.4% | 243.65 | C10H10ClNO4 | 100 MG
SDP

Catalog No. 5000300080
Encompass_Preferred

4-Acetamido-5-chloro-2-methoxybenzoic acid is a synthetic drug intermediate (Metoclopramide Impurity 12) used in the synthesis of active pharmaceutical compounds. It is supplied as a high-purity solid intended for research, analytical method development, and impurity profiling.

  • High purity: 99.4% suitable for analytical and synthetic applications.
  • Molecular weight: 243.65 g/mol.
  • Appearance: solid.
  • Useful as a drug intermediate and for impurity characterization studies.
  • Available in small pack sizes for research and development.
  • SDS available for handling and safety information.

Catalog No. 50-003-00080 Supplier Medchemexpress LLC Supplier No. HYW036175100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Acetamido-5-chloro-2-methoxybenzoic acid is a synthetic drug intermediate (Metoclopramide Impurity 12) used in the synthesis of active pharmaceutical compounds. It is supplied as a high-purity solid intended for research, analytical method development, and impurity profiling.

  • High purity: 99.4% suitable for analytical and synthetic applications.
  • Molecular weight: 243.65 g/mol.
  • Appearance: solid.
  • Useful as a drug intermediate and for impurity characterization studies.
  • Available in small pack sizes for research and development.
  • SDS available for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.