Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-acetamido-5-chloro-2-methoxybenzoic acid | 24201-13-6 | MFCD07783714 | 99.4% | 243.65 | C10H10ClNO4 | 1 G
SDP

Catalog No. 5000299560
Encompass_Preferred

4-Acetamido-5-chloro-2-methoxybenzoic acid is a benzoic-acid derivative used as a drug intermediate and analytical impurity reference in medicinal chemistry and process development. It is supplied for research use where high chemical purity and traceable analytical documentation are required.

  • High purity (99.39% by LCMS).
  • Molecular formula C10H10ClNO4.
  • Molecular weight 243.65 g·mol⁻¹.
  • Suitable as a drug intermediate and impurity reference standard.
  • Supplied with certificate of analysis and safety data sheet for traceability.

Catalog No. 50-002-99560 Supplier Medchemexpress LLC Supplier No. HYW0361751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Acetamido-5-chloro-2-methoxybenzoic acid is a benzoic-acid derivative used as a drug intermediate and analytical impurity reference in medicinal chemistry and process development. It is supplied for research use where high chemical purity and traceable analytical documentation are required.

  • High purity (99.39% by LCMS).
  • Molecular formula C10H10ClNO4.
  • Molecular weight 243.65 g·mol⁻¹.
  • Suitable as a drug intermediate and impurity reference standard.
  • Supplied with certificate of analysis and safety data sheet for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.