Learn More
Medchemexpress LLC 4-acetamido-5-chloro-2-methoxybenzoic acid | 24201-13-6 | MFCD07783714 | 99.4% | 243.65 | C10H10ClNO4 | 250 MG
SDP
Catalog No. 5000303428
Encompass_Preferred
4-Acetamido-5-chloro-2-methoxybenzoic acid is a benzoic acid derivative used as a drug intermediate and analytical reference (identified as Metoclopramide Impurity 12). It is supplied as a white to off-white solid and is characterized by LCMS and 1H NMR, with documented stability and recommended storage conditions for powder and solutions.
  • High purity suitable for analytical and synthetic applications.
  • Documented analytical data for identity confirmation.
  • Clear storage guidance to maintain sample stability.
  • Defined molecular properties to support accurate synthesis and formulation.
Catalog No. 50-003-03428 Supplier Medchemexpress LLC Supplier No. 250MGHYW036175
4-Acetamido-5-chloro-2-methoxybenzoic acid is a benzoic acid derivative used as a drug intermediate and analytical reference (identified as Metoclopramide Impurity 12). It is supplied as a white to off-white solid and is characterized by LCMS and 1H NMR, with documented stability and recommended storage conditions for powder and solutions.
  • High purity suitable for analytical and synthetic applications.
  • Documented analytical data for identity confirmation.
  • Clear storage guidance to maintain sample stability.
  • Defined molecular properties to support accurate synthesis and formulation.