Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-acetamido-5-chloro-2-methoxybenzoic acid | 24201-13-6 | MFCD07783714 | 99.4% | 243.65 g/mol | C10H10ClNO4 | 500 MG
SDP

Catalog No. 5000299404
Encompass_Preferred

4-Acetamido-5-chloro-2-methoxybenzoic acid is a chemical intermediate used in pharmaceutical research and synthesis; it is also identified as Metoclopramide Impurity 12 and is supplied as a white to off-white solid suitable for medicinal chemistry applications.

  • High purity: 99.4%.
  • Molecular formula: C10H10ClNO4; molecular weight: 243.65 g/mol.
  • Appearance: white to off-white solid.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Packaging: available in sizes from 100 mg to 10 g.

Catalog No. 50-002-99404 Supplier Medchemexpress LLC Supplier No. HYW036175500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Acetamido-5-chloro-2-methoxybenzoic acid is a chemical intermediate used in pharmaceutical research and synthesis; it is also identified as Metoclopramide Impurity 12 and is supplied as a white to off-white solid suitable for medicinal chemistry applications.

  • High purity: 99.4%.
  • Molecular formula: C10H10ClNO4; molecular weight: 243.65 g/mol.
  • Appearance: white to off-white solid.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Packaging: available in sizes from 100 mg to 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.