Learn More
AdipoGen 4-Acetamido-5-nitrophthalic anhydride (1 g)
SDP
Supplier:  AdipoGen CDXA0245G001
Encompass_Preferred
Chemical. CAS: 209324-72-1. Formula: C10H6N2O6. MW: 250.16. Building Block/Intermediate used for the synthesis of diaminofluorescein derivatives.
Catalog No. 50-205-4261