Learn More
Medchemexpress LLC 4-acetamido-N-(tert-butyl)benzamide | 183619-38-7 | 99.8% | 234.29 | C13H18N2O2 | 10 MG
SDP
Catalog No. 5000213778
Encompass_Preferred
CPI-1189 is a small-molecule TNF-α release inhibitor used in preclinical research to investigate inflammatory and neuroprotective mechanisms. It has been reported to inhibit p38 phosphorylation and reduce apoptosis in cellular models, making it useful for studies of neurodegeneration, HIV-related neuronal injury, and oxidative stress.
  • Orally active TNF-α release inhibitor suitable for cell-based assays.
  • Modulates p38 signaling to reduce inflammation-related apoptosis.
  • High chemical purity for reproducible experimental results.
  • Available in small-scale quantities for research applications.
  • Supplied with datasheet listing CAS, molecular weight, and structural identifiers.
Catalog No. 50-002-13778 Supplier Medchemexpress LLC Supplier No. HY10037610MG
CPI-1189 is a small-molecule TNF-α release inhibitor used in preclinical research to investigate inflammatory and neuroprotective mechanisms. It has been reported to inhibit p38 phosphorylation and reduce apoptosis in cellular models, making it useful for studies of neurodegeneration, HIV-related neuronal injury, and oxidative stress.
  • Orally active TNF-α release inhibitor suitable for cell-based assays.
  • Modulates p38 signaling to reduce inflammation-related apoptosis.
  • High chemical purity for reproducible experimental results.
  • Available in small-scale quantities for research applications.
  • Supplied with datasheet listing CAS, molecular weight, and structural identifiers.