Learn More
Medchemexpress LLC 4-acetamido-N-(tert-butyl)benzamide | 183619-38-7 | MFCD00774476 | 99.8% | 234.29 g/mol | C13H18N2O2 | 25 MG
SDP
Catalog No. 5000213580
Encompass_Preferred
CPI-1189 is a small-molecule, orally active inhibitor of tumor necrosis factor-alpha (TNF-α) release that inhibits p38 MAPK phosphorylation and can prevent apoptosis. It is used as a research tool in studies of neuroprotection, inflammation, HIV-related pathology, and neurodegenerative disease models.
  • Orally active TNF-α release inhibitor.
  • Inhibits p38 phosphorylation to modulate inflammatory signaling.
  • Demonstrated neuroprotective and antiapoptotic effects in cellular models.
  • High chemical purity suitable for biological research.
  • Solid, white to off-white appearance for easy handling.
  • Supplied in research-scale quantities for laboratory use.
Catalog No. 50-002-13580 Supplier Medchemexpress LLC Supplier No. HY10037625MG
CPI-1189 is a small-molecule, orally active inhibitor of tumor necrosis factor-alpha (TNF-α) release that inhibits p38 MAPK phosphorylation and can prevent apoptosis. It is used as a research tool in studies of neuroprotection, inflammation, HIV-related pathology, and neurodegenerative disease models.
  • Orally active TNF-α release inhibitor.
  • Inhibits p38 phosphorylation to modulate inflammatory signaling.
  • Demonstrated neuroprotective and antiapoptotic effects in cellular models.
  • High chemical purity suitable for biological research.
  • Solid, white to off-white appearance for easy handling.
  • Supplied in research-scale quantities for laboratory use.