Learn More
Cayman Chemical 4-Acetamido Antipyrine, Purity: ≥95%, CAS: 83-15-8, 250 mg
SDP
Catalog No. 502942425
Encompass_Preferred
An active metabolite of metamizole; inhibits β-galactosidase activity, a marker of LXRα activity, in a yeast two-hybrid assay (IC20 = 79.9 µM); has been found as a micropollutant in reclaimed wastewater
Catalog No. 50-294-2425 Supplier Cayman Chemical Supplier No. 40308250 MG
An active metabolite of metamizole; inhibits β-galactosidase activity, a marker of LXRα activity, in a yeast two-hybrid assay (IC20 = 79.9 µM); has been found as a micropollutant in reclaimed wastewater