Learn More
Cayman Chemical 4-AcetamIdo ntIpyrIn 25mg
SDP
Catalog No. 502945952
Encompass_Preferred
An active metabolite of metamizole; inhibits β-galactosidase activity, a marker of LXRα activity, in a yeast two-hybrid assay (IC20 = 79.9 µM); has been found as a micropollutant in reclaimed wastewater
Catalog No. 50-294-5952 Supplier Cayman Chemical Supplier No. 4030825 MG
An active metabolite of metamizole; inhibits β-galactosidase activity, a marker of LXRα activity, in a yeast two-hybrid assay (IC20 = 79.9 µM); has been found as a micropollutant in reclaimed wastewater