Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. A011525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A011525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A011525G Supplier TCI America Supplier No. A011525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121-61-9
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
MDL Number MFCD00035784
InChI Key PKOFBDHYTMYVGJ-UHFFFAOYSA-N
Synonym N4-Acetylsulfanilamide
PubChem CID 8482
IUPAC Name N-(4-sulfamoylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 25 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidobenzenesulfonamide
EINECSNumber (3)-1989
RTECSNumber AE7015000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.