Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzenethiol 90.0+%, TCI America™
SDP

Catalog No. A19095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19095G 5 g
A190925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19095G Supplier TCI America Supplier No. A19095G

Chemical Identifiers

CAS 1126-81-4
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.23
MDL Number MFCD00004848
InChI Key AYEQJLOHMLYKAV-UHFFFAOYSA-N
Synonym 4-acetamidothiophenol, 4'-mercaptoacetanilide, 4-acetamidobenzenethiol, n-4-sulfanylphenyl acetamide, 4-acetamido thiophenol, n-4-mercaptophenyl acetamide, acetanilide, 4'-mercapto, 4-acetaminothiophenol, 4-mercaptoacetanilide, 4-acetylaminothiophenol
PubChem CID 70777
IUPAC Name N-(4-sulfanylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(S)C=C1

Specifications

Melting Point 153°C
Color Yellow
Quantity 5 g
Formula Weight 167.23
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.