Learn More
Medchemexpress LLC 4-acetamidobutanoic acid-d3 (N-acetyl GABA-d3) | 00-00-0 | MFCD00020552 | 148.17 g/mol | C6H8D3NO3 | 1 MG
SDP
Catalog No. 5000245415
Encompass_Preferred
4-acetamidobutanoic acid-d3 (N-acetyl GABA-d3) is the deuterated analog of 4-acetamidobutanoic acid, supplied for use as an isotopically labeled internal standard in quantitative analytical workflows. Formula C6H8D3NO3, molecular weight 148.17 g/mol. Supplied in milligram quantities for use in NMR, GC-MS, and LC-MS.
  • Isotopically labeled internal standard for quantitative analysis.
  • Provides a clear mass shift for accurate LC-MS and GC-MS quantitation.
  • Compatible with NMR for deuterium-based tracing and structural studies.
  • Supplied in small, precisely measured amounts for analytical assays.
  • Useful as a tracer in metabolic and pharmacokinetic investigations.
Catalog No. 50-002-45415 Supplier Medchemexpress LLC Supplier No. HY101411S1MG
4-acetamidobutanoic acid-d3 (N-acetyl GABA-d3) is the deuterated analog of 4-acetamidobutanoic acid, supplied for use as an isotopically labeled internal standard in quantitative analytical workflows. Formula C6H8D3NO3, molecular weight 148.17 g/mol. Supplied in milligram quantities for use in NMR, GC-MS, and LC-MS.
  • Isotopically labeled internal standard for quantitative analysis.
  • Provides a clear mass shift for accurate LC-MS and GC-MS quantitation.
  • Compatible with NMR for deuterium-based tracing and structural studies.
  • Supplied in small, precisely measured amounts for analytical assays.
  • Useful as a tracer in metabolic and pharmacokinetic investigations.