Learn More
Cayman Chemical 4-AcetamIdobutyrIc AcId 10g
SDP
Catalog No. 502990279
Encompass_Preferred
A GABA derivative; urinary levels are elevated in patients with stage 5 chronic kidney disease compared to patients stage 3 kidney disease and those without kidney disease
Catalog No. 50-299-0279 Supplier Cayman Chemical Supplier No. 2988110 G
Add to Cart
Add to Cart
A GABA derivative; urinary levels are elevated in patients with stage 5 chronic kidney disease compared to patients stage 3 kidney disease and those without kidney disease