Learn More
Medchemexpress LLC 4-Acetamidophenyl acetate | 2623-33-8 | >=97.0% | C10H11NO3 | 1 ML
SDP
Catalog No. 5000434143
Encompass_Preferred
4-Acetamidophenyl acetate is an impurity of Acetaminophen and acts as an intermediate in its synthesis. It is classified as harmful if swallowed, causes skin irritation, causes serious eye irritation, and may cause respiratory irritation.
  • Selective COX-2 inhibitor (IC50=25.8 μM)
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Undergoes hydrolytic cleavage independent of microsomes and cofactors in rat liver microsomal metabolism assays in vitro
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-004-34143 Supplier Medchemexpress LLC Supplier No. HY6600410MM/1ML
4-Acetamidophenyl acetate is an impurity of Acetaminophen and acts as an intermediate in its synthesis. It is classified as harmful if swallowed, causes skin irritation, causes serious eye irritation, and may cause respiratory irritation.
  • Selective COX-2 inhibitor (IC50=25.8 μM)
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Undergoes hydrolytic cleavage independent of microsomes and cofactors in rat liver microsomal metabolism assays in vitro
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation