Learn More
Medchemexpress LLC 4-Acetamidophenyl acetate | 2623-33-8 | 97.0% | C10H11NO3 | 25 G
SDP
Catalog No. 5000358947
Encompass_Preferred
4-Acetamidophenyl acetate is an impurity of Acetaminophen and acts as an intermediate in its synthesis. Acetaminophen is described as a selective COX-2 inhibitor and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. This product is for research use only.
  • Impurity of Acetaminophen
  • Intermediate in the synthesis of Acetaminophen
  • Available in solid form (white to off-white)
  • Solubility in DMSO: 100 mg/mL (517.60 mM)
  • In vivo dissolution methods are provided
  • Hydrolytic cleavage is independent of microsomes and cofactors in rat liver microsomal metabolism assays
Catalog No. 50-003-58947 Supplier Medchemexpress LLC Supplier No. 25GHY66004
4-Acetamidophenyl acetate is an impurity of Acetaminophen and acts as an intermediate in its synthesis. Acetaminophen is described as a selective COX-2 inhibitor and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. This product is for research use only.
  • Impurity of Acetaminophen
  • Intermediate in the synthesis of Acetaminophen
  • Available in solid form (white to off-white)
  • Solubility in DMSO: 100 mg/mL (517.60 mM)
  • In vivo dissolution methods are provided
  • Hydrolytic cleavage is independent of microsomes and cofactors in rat liver microsomal metabolism assays