Learn More
Medchemexpress LLC 4-Acetamidophenyl acetate | 2623-33-8 | 97.0% | C10H11NO3 | 5 G
SDP
Catalog No. 5000434451
Encompass_Preferred
4-Acetamidophenyl acetate is an impurity of Acetaminophen and an intermediate in its synthesis. Acetaminophen is known as a selective COX-2 inhibitor (IC50=25.8 μM) and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. In in vitro rat liver microsomal metabolism assays, this compound's hydrolytic cleavage over 90 minutes was observed to be independent of microsomes and cofactors.
  • Impurity of acetaminophen
  • Intermediate in acetaminophen synthesis
  • Hydrolytic cleavage independent of microsomes and cofactors in rat liver microsomal metabolism assays in vitro
  • For research use only
Catalog No. 50-004-34451 Supplier Medchemexpress LLC Supplier No. HY660045G
Add to Cart
Add to Cart
4-Acetamidophenyl acetate is an impurity of Acetaminophen and an intermediate in its synthesis. Acetaminophen is known as a selective COX-2 inhibitor (IC50=25.8 μM) and a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. In in vitro rat liver microsomal metabolism assays, this compound's hydrolytic cleavage over 90 minutes was observed to be independent of microsomes and cofactors.
  • Impurity of acetaminophen
  • Intermediate in acetaminophen synthesis
  • Hydrolytic cleavage independent of microsomes and cofactors in rat liver microsomal metabolism assays in vitro
  • For research use only