Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidopyridine 98.0+%, TCI America™
SDP

Catalog No. A18835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18835G 5 g
A188325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18835G Supplier TCI America Supplier No. A18835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5221-42-1
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
InChI Key BIJAWQUBRNHZGE-UHFFFAOYSA-N
Synonym N-(4-Pyridyl)acetamide
PubChem CID 21288
IUPAC Name N-pyridin-4-ylacetamide
SMILES CC(=O)NC1=CC=NC=C1

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidopyridine
RTECSNumber AC7875000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.