Learn More
Cayman Chemical 4-acetoxy DiPT (acetate), Purity: ≥95%, CAS: 2749435-22-9, 10 mg
SDP
Catalog No. 502951498
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces the HTR in mice, indicating hallucinogenic potential; substitutes for DOM in a two-lever drug discrimination test; has a high potential for abuse; intended for research and forensic applications
Catalog No. 50-295-1498 Supplier Cayman Chemical Supplier No. 1131110 MG
An analytical reference standard categorized as a tryptamine; induces the HTR in mice, indicating hallucinogenic potential; substitutes for DOM in a two-lever drug discrimination test; has a high potential for abuse; intended for research and forensic applications