Learn More
Cayman Chemical 4-acetoxy MALT 1mg
SDP
Catalog No. 502937173
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; intended for research and forensic applications
Catalog No. 50-293-7173 Supplier Cayman Chemical Supplier No. 272561 MG
An analytical reference standard categorized as a tryptamine; intended for research and forensic applications