Learn More
Medchemexpress LLC 4-Acetoxycinnamic ac 100mg
SDP
Catalog No. 5000206723
Encompass_Preferred
4-Acetoxycinnamic acid is an acetate ester obtained by the formal condensation of the hydroxy group of trans-4-coumaric acid with acetic acid 4-Acetoxycinnamic acid is a member of cinnamic acids and a member of phenyl acetates 4-Acetoxycinnamic acid derives from a trans-4-coumaric acid[1]
Catalog No. 50-002-06723 Supplier Medchemexpress LLC Supplier No. HYW102375100MG
4-Acetoxycinnamic acid is an acetate ester obtained by the formal condensation of the hydroxy group of trans-4-coumaric acid with acetic acid 4-Acetoxycinnamic acid is a member of cinnamic acids and a member of phenyl acetates 4-Acetoxycinnamic acid derives from a trans-4-coumaric acid[1]