Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-acetyl-2-methylbenzoic acid | 55860-35-0 | MFCD20639525 | 99.9% | 178.19 | C10H10O3 | 25 G
SDP

Catalog No. 5000208836
Encompass_Preferred

Fluralaner intermediate (4-acetyl-2-methylbenzoic acid) is a chemical intermediate used in the synthesis of fluralaner and related active pharmaceutical compounds. It is supplied at high purity for research and synthetic applications.

  • Chemical name: 4-acetyl-2-methylbenzoic acid.
  • Cas number: 55860-35-0.
  • Molecular formula: C10H10O3.
  • Molecular weight: 178.19.
  • Purity: 99.9%.
  • Available in multiple package sizes, including 25 g.
  • Intended for research and synthetic applications.

Catalog No. 50-002-08836 Supplier Medchemexpress LLC Supplier No. 25GHYW076700
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluralaner intermediate (4-acetyl-2-methylbenzoic acid) is a chemical intermediate used in the synthesis of fluralaner and related active pharmaceutical compounds. It is supplied at high purity for research and synthetic applications.

  • Chemical name: 4-acetyl-2-methylbenzoic acid.
  • Cas number: 55860-35-0.
  • Molecular formula: C10H10O3.
  • Molecular weight: 178.19.
  • Purity: 99.9%.
  • Available in multiple package sizes, including 25 g.
  • Intended for research and synthetic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.