Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetyl-4'-methylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. A13225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13225G Supplier TCI America Supplier No. A13225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5748-38-9
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00191374
InChI Key GNIQQKORSMFYPE-UHFFFAOYSA-N
Synonym 4-Acetyl-4′C-methyldiphenyl
PubChem CID 4301905
IUPAC Name 1-[4-(4-methylphenyl)phenyl]ethanone
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetyl-4′-methylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.