Learn More
Medchemexpress LLC 4'-acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1,1'-biphenyl]-4-carboxamide | 162408-66-4 | 99.6% | 485.62 | C30H35N3O3 | 100 MG
SDP
Catalog No. 5000213212
Encompass_Preferred
A potent, selective dopamine D3 receptor antagonist used for receptor pharmacology research. It shows sub-nanomolar affinity for the D3 receptor and strong selectivity versus related dopamine subtypes.
  • Potent dopamine D3 receptor antagonist with Ki 0.4 nM.
  • Greater than 100-fold selectivity for D3 over D4 and D1.
  • High purity (batch COA: 99.6%).
  • White to off-white solid powder; molecular formula C30H35N3O3; molecular weight 485.62.
  • Suitable for in vitro and in vivo receptor pharmacology studies.
  • Store powder at -20°C; in solution store at -80°C for long-term stability.
Catalog No. 50-002-13212 Supplier Medchemexpress LLC Supplier No. HY101382100MG
A potent, selective dopamine D3 receptor antagonist used for receptor pharmacology research. It shows sub-nanomolar affinity for the D3 receptor and strong selectivity versus related dopamine subtypes.
  • Potent dopamine D3 receptor antagonist with Ki 0.4 nM.
  • Greater than 100-fold selectivity for D3 over D4 and D1.
  • High purity (batch COA: 99.6%).
  • White to off-white solid powder; molecular formula C30H35N3O3; molecular weight 485.62.
  • Suitable for in vitro and in vivo receptor pharmacology studies.
  • Store powder at -20°C; in solution store at -80°C for long-term stability.